Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl27
Entrez Gene ID
100759321
Gene Name
mitochondrial ribosomal protein L27
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613846: 1746643 - 1752042 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501355.2 XP_003501403.1 3 - 4 260 72
GenBank mRNA JI886442.1 - 3 - 4 263 72
JP063999.1 - 3 - 4 263 72

View Full Product Details