Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm12
Entrez Gene ID
100761986
Gene Name
PR domain containing 12
Gene Aliases
I79_022053
Chromosome Location
Chr.Un|NW_003615743: 45671 - 60194 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513122.1 XP_003513170.1 4 - 5 681 77

View Full Product Details