Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abra
Entrez Gene ID
100767760
Gene Name
actin-binding Rho activating protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613897: 1497098 - 1502475 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502430.1 XP_003502478.1 1 - 2 656 63

View Full Product Details