Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc22a20
Entrez Gene ID
100758921
Gene Name
solute carrier family 22, member 20
Gene Aliases
I79_024884
Chromosome Location
Chr.Un|NW_003617890: 43221 - 49320 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515375.1 XP_003515423.1 4 - 5 800 68

View Full Product Details