Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlx4
Entrez Gene ID
100757892
Gene Name
distal-less homeobox 4
Gene Aliases
I79_010905
Chromosome Location
Chr.Un|NW_003614015: 998981 - 1008670 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644482.1 XP_007642672.1 5 - 6 2015 105

View Full Product Details