Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hpcal4
Entrez Gene ID
100772357
Gene Name
hippocalcin like 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614116: 800432 - 803478 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645567.1 XP_007643757.1 1 - 2 159 74

View Full Product Details