Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snrpd3
Entrez Gene ID
100768269
Gene Name
small nuclear ribonucleoprotein D3 polypeptide 18kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614478: 197840 - 214800 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648727.1 XP_007646917.1 2 - 3 709 69

View Full Product Details