Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Steap3
Entrez Gene ID
100761057
Gene Name
STEAP family member 3, metalloreductase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613737: 1551978 - 1575456 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499444.2 XP_003499492.1 4 - 5 1297 66

View Full Product Details