Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kctd13
Entrez Gene ID
100753748
Gene Name
potassium channel tetramerization domain containing 13
Gene Aliases
I79_018365
Chromosome Location
Chr.Un|NW_003614830: 231930 - 246092 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510128.2 XP_003510176.1 3 - 4 561 118

View Full Product Details