Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd30a
Entrez Gene ID
100769687
Gene Name
ankyrin repeat domain 30A
Gene Aliases
Ankrd26
Chromosome Location
Chr.Un|NW_003614048: 36695 - 98161 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504266.2 XP_003504314.1 32 - 33 5106 100
XM_007644818.1 XP_007643008.1 32 - 33 5121 100

View Full Product Details