Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nr1d2
Entrez Gene ID
100768895
Gene Name
nuclear receptor subfamily 1, group D, member 2
Gene Aliases
I79_003980
Chromosome Location
Chr.Un|NW_003613684: 2444087 - 2466864 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498212.2 XP_003498260.1 5 - 6 1312 91

View Full Product Details