Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syce2
Entrez Gene ID
103161434
Gene Name
synaptonemal complex central element protein 2
Gene Aliases
I79_008344
Chromosome Location
Chr.Un|NW_003613865: 349758 - 355394 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642539.1 XP_007640729.1 2 - 3 292 104

View Full Product Details