Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map9
Entrez Gene ID
100773111
Gene Name
microtubule-associated protein 9
Gene Aliases
I79_005564, Mtap9
Chromosome Location
Chr.Un|NW_003613739: 510586 - 542833 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499480.2 XP_003499528.1 13 - 14 2085 66
XM_007640219.1 XP_007638409.1 13 - 14 2119 66

View Full Product Details