Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rabif
Entrez Gene ID
100753301
Gene Name
RAB interacting factor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613706: 345286 - 361496 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498850.2 XP_003498898.1 1 - 2 155 63

View Full Product Details