Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vsig2
Entrez Gene ID
100769642
Gene Name
V-set and immunoglobulin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616891: 71413 - 77362 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514727.2 XP_003514775.1 2 - 3 526 84

View Full Product Details