Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slx1a
Entrez Gene ID
100752848
Gene Name
SLX1 structure-specific endonuclease subunit homolog A (S. cerevisiae)
Gene Aliases
I79_018346
Chromosome Location
Chr.Un|NW_003614830: 54 - 2193 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654886.1 XP_007653076.1 1 - 2 333 101

View Full Product Details