Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mixl1
Entrez Gene ID
100774111
Gene Name
Mix paired-like homeobox
Gene Aliases
I79_014847
Chromosome Location
Chr.Un|NW_003614340: 263680 - 269366 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647800.1 XP_007645990.1 4 - 5 432 98

View Full Product Details