Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhl33
Entrez Gene ID
100754250
Gene Name
kelch-like family member 33
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614668: 262530 - 270146 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509205.2 XP_003509253.2 3 - 4 1954 104

View Full Product Details