Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf39
Entrez Gene ID
100753435
Gene Name
ring finger protein 39
Gene Aliases
I79_015092
Chromosome Location
Chr.Un|NW_003614381: 364541 - 369303 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648026.1 XP_007646216.1 3 - 4 318 71

View Full Product Details