Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxl3
Entrez Gene ID
100761851
Gene Name
F-box and leucine-rich repeat protein 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613975: 965536 - 981983 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503484.2 XP_003503532.1 3 - 4 639 66
GenBank mRNA JI879404.1 - 2 - 3 466 66

View Full Product Details