Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cbx4
Entrez Gene ID
100756554
Gene Name
chromobox homolog 4
Gene Aliases
I79_014166
Chromosome Location
Chr.Un|NW_003614274: 608638 - 619947 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647241.1 XP_007645431.1 9 - 10 1141 50

View Full Product Details