Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdnf
Entrez Gene ID
100751621
Gene Name
cerebral dopamine neurotrophic factor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613621: 3678587 - 3695923 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496417.2 XP_003496465.1 3 - 4 565 77
GenBank mRNA JI863265.1 - 3 - 4 486 77
JP050650.1 - 3 - 4 486 77

View Full Product Details