Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptx4
Entrez Gene ID
100774180
Gene Name
pentraxin 4, long
Gene Aliases
I79_007929
Chromosome Location
Chr.Un|NW_003613852: 1153144 - 1158308 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501480.2 XP_003501528.1 1 - 2 685 85

View Full Product Details