Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dusp27
Entrez Gene ID
100768141
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613889: 1355889 - 1384702 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502287.1 XP_003502335.1 4 - 5 658 65

View Full Product Details