Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dolk
Entrez Gene ID
100768385
Gene Name
dolichol kinase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616251: 29951 - 32035 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513968.2 XP_003514016.1 1 - 1 1518 67

View Full Product Details