Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyyr1
Entrez Gene ID
100757225
Gene Name
cysteine/tyrosine-rich 1
Gene Aliases
I79_011925
Chromosome Location
Chr.Un|NW_003614089: 6017 - 94335 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504813.1 XP_003504861.1 1 - 2 76 76

View Full Product Details