Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnfsf9
Entrez Gene ID
100750679
Gene Name
tumor necrosis factor (ligand) superfamily, member 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613861: 1288844 - 1291061 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501810.2 XP_003501858.1 1 - 2 464 75

View Full Product Details