Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cylc2
Entrez Gene ID
100763548
Gene Name
cylicin, basic protein of sperm head cytoskeleton 2
Gene Aliases
I79_001126
Chromosome Location
Chr.Un|NW_003613603: 762928 - 771055 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495863.1 XP_003495911.1 2 - 3 60 92

View Full Product Details