Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam98c
Entrez Gene ID
100765434
Gene Name
family with sequence similarity 98, member C
Gene Aliases
I79_010747
Chromosome Location
Chr.Un|NW_003614005: 466689 - 474644 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503834.2 XP_003503882.1 4 - 5 465 76

View Full Product Details