Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Palm3
Entrez Gene ID
100768138
Gene Name
paralemmin 3
Gene Aliases
I79_008380
Chromosome Location
Chr.Un|NW_003613865: 1253765 - 1262168 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501936.2 XP_003501984.1 6 - 7 457 63

View Full Product Details