Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Steap4
Entrez Gene ID
100750411
Gene Name
STEAP family member 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614224: 68143 - 112378 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506277.2 XP_003506325.1 4 - 5 1193 69

View Full Product Details