Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim16
Entrez Gene ID
100753576
Gene Name
tripartite motif containing 16
Gene Aliases
I79_001756
Chromosome Location
Chr.Un|NW_003613619: 2450151 - 2469383 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496345.2 XP_003496393.1 5 - 6 1463 75

View Full Product Details