Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kbtbd12
Entrez Gene ID
100766790
Gene Name
kelch repeat and BTB (POZ) domain containing 12
Gene Aliases
I79_006448
Chromosome Location
Chr.Un|NW_003613782: 14869 - 72231 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500219.2 XP_003500267.1 1 - 2 1090 83

View Full Product Details