Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxa10
Entrez Gene ID
100762719
Gene Name
homeobox A10
Gene Aliases
I79_008458
Chromosome Location
Chr.Un|NW_003613870: 83673 - 87214 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642618.1 XP_007640808.1 2 - 3 180 68

View Full Product Details