Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc26
Entrez Gene ID
100771542
Gene Name
leucine rich repeat containing 26
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616080: 46332 - 47415 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513697.1 XP_003513745.1 1 - 2 687 57

View Full Product Details