Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cideb
Entrez Gene ID
100769570
Gene Name
cell death-inducing DFFA-like effector b
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613875: 972732 - 977359 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502087.2 XP_003502135.2 5 - 6 647 72

View Full Product Details