Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam50b
Entrez Gene ID
100759390
Gene Name
family with sequence similarity 50, member B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613650: 631516 - 632523 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007612653.1 XP_007610843.1 1 - 1 723 77

View Full Product Details