Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Irs4
Entrez Gene ID
100773159
Gene Name
insulin receptor substrate 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614518: 486135 - 509412 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648937.1 XP_007647127.1 1 - 2 107 95

View Full Product Details