Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slamf8
Entrez Gene ID
100771668
Gene Name
SLAM family member 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613783: 1069942 - 1077866 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500232.1 XP_003500280.1 3 - 4 668 62

View Full Product Details