Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC100761903
Entrez Gene ID
100761903
Gene Name
Uncharacterized protein C1orf31-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613636: 600517 - 602811 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496801.2 XP_003496849.1 1 - 2 387 111

View Full Product Details