Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrtm2
Entrez Gene ID
100752413
Gene Name
leucine-rich repeats and transmembrane domains 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613787: 738566 - 745139 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500322.1 XP_003500370.1 2 - 3 659 86

View Full Product Details