Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hspb6
Entrez Gene ID
100774774
Gene Name
heat shock protein, alpha-crystallin-related, B6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614167: 675924 - 676440 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505571.2 XP_003505619.2 1 - 2 162 58

View Full Product Details