Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clrn1
Entrez Gene ID
100768820
Gene Name
clarin 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613922: 795920 - 833052 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502877.2 XP_003502925.1 1 - 2 248 86

View Full Product Details