Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Igfbpl1
Entrez Gene ID
100769562
Gene Name
insulin-like growth factor binding protein-like 1
Gene Aliases
I79_006278
Chromosome Location
Chr.Un|NW_003613773: 445430 - 459280 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500098.1 XP_003500146.1 1 - 2 58 76

View Full Product Details