Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lgals7
Entrez Gene ID
100767471
Gene Name
lectin, galactoside-binding, soluble, 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614005: 770440 - 778542 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503837.1 XP_003503885.1 1 - 2 148 76

View Full Product Details