Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spsb2
Entrez Gene ID
100757837
Gene Name
splA/ryanodine receptor domain and SOCS box containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613610: 3911590 - 3913073 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496065.2 XP_003496113.2 1 - 2 829 60

View Full Product Details