Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tomm5
Entrez Gene ID
100751863
Gene Name
translocase of outer mitochondrial membrane 5 homolog (yeast)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614087: 936407 - 939097 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504797.2 XP_003504845.1 1 - 2 222 84

View Full Product Details