Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exosc5
Entrez Gene ID
100773940
Gene Name
exosome component 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615239: 192907 - 214764 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652329.1 XP_007650519.1 4 - 5 574 83

View Full Product Details