Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab19
Entrez Gene ID
100770627
Gene Name
RAB19, member RAS oncogene family
Gene Aliases
I79_009733
Chromosome Location
Chr.Un|NW_003613933: 453906 - 458505 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503019.2 XP_003503067.1 2 - 3 385 71

View Full Product Details