Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppm1a
Entrez Gene ID
100773654
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615418: 87620 - 125838 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512313.2 XP_003512361.2 5 - 6 1156 98
XM_007652828.1 XP_007651018.1 5 - 6 1192 98
GenBank mRNA JI895092.1 - 4 - 5 1130 98
JP058612.1 - 4 - 5 1130 98

View Full Product Details